missing translation for 'onlineSavingsMsg'
Learn More

2-Methylsulphonylbenzoic acid, 95%, Thermo Scientific™

Product Code. 10656641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10656641 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10656641 Supplier Thermo Scientific Acros Supplier No. 424830025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 33963-55-2
CAS Min % 94.0
CAS Max % 100.0
Melting Point 136.0°C to 141.0°C
Color Green or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C8H8O4S
Linear Formula SO2CH3C6H4COOH
MDL Number MFCD00216492
Quantity 2.5 g
Synonym 2-methylsulfonyl benzoic acid, 2-methanesulfonylbenzoic acid, 2-methanesulfonyl-benzoic acid, 2-methanesulfonyl benzoic acid, benzoic acid, 2-methylsulfonyl, 2-methylsulfonyl benzenecarboxylic acid, 2-methylsulphonyl benzoic acid, 2-mesylbenzoic acid, pubchem18527, methylsulfonylbenzoic acid
Solubility Information Solubility in water: negligible
InChI Key BZSXEZOLBIJVQK-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=CC=C1C(=O)O
IUPAC Name 2-methylsulfonylbenzoic acid
Molecular Weight (g/mol) 200.22
PubChem CID 284342
Formula Weight 200.22
Percent Purity 95%
Physical Form Chunks and/or Powder
Chemical Name or Material 2-Methylsulphonylbenzoic acid, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylsulphonylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.