missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfonyl)phenol, 98%

Product Code. 15315048 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15315048 5 g 5g
15305048 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15315048 Supplier Thermo Scientific Acros Supplier No. 455750050
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27489-33-4 | C7H8O3S | 172.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27489-33-4
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.2
InChI Key ZWTZHHHSRDUVRT-UHFFFAOYSA-N
PubChem CID 598998
IUPAC Name 2-methylsulfonylphenol
SMILES CS(=O)(=O)C1=CC=CC=C1O

Specifications

Melting Point 88.0°C to 90.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Linear Formula HOC6H4SO2CH3
Quantity 5 g
Synonym 2-methylsulfonyl phenol, 2-methanesulfonylphenol, 2-methylsulfonyl benzenol, phenol, 2-methylsulfonyl, phenol,2-methylsulfonyl, phenol, o-methylsulfonyl, methylsulfonylphenol, methylsulfonylbenzenol, acmc-1cbus, 2-methanesulfonyl-phenol
Formula Weight 172.2
Percent Purity 98%
Chemical Name or Material 2-(Methylsulfonyl)phenol
compliance-icons
Product Identifier
  • 2-(Methylsulfonyl)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.