missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfonyl)ethanol, 97%

Product Code. 15408819
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408819 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15408819 Supplier Thermo Scientific Alfa Aesar Supplier No. L03159.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15205-66-0 | C3H8O3S | 124.154 g/mol

2-(Methylsulfonyl)ethanol may be used as reagent in the synthesis of phenols from aryl fluorides. It may be used in the preparation of 2-(methylsulfonyl)ethyl N,N-bis(2-chloroethyl)phosphorodiamidate. 2-Hydroxyethyl Methyl Sulfone, is the versatile building block used in the synthesis of more complex sulfur containing compounds. It can be used for the studies of inhibitor and substrate moieties to acetylcholinesterase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylsulfonyl)ethanol may be used as reagent in the synthesis of phenols from aryl fluorides. It may be used in the preparation of 2-(methylsulfonyl)ethyl N,N-bis(2-chloroethyl)phosphorodiamidate. 2-Hydroxyethyl Methyl Sulfone,
TRUSTED_SUSTAINABILITY

Specifications

CAS 15205-66-0
Melting Point 27°C to 30°C
Boiling Point 148°C to 149°C (1 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C3H8O3S
Refractive Index 1.479
MDL Number MFCD00014747
Quantity 5 g
Beilstein 1746915
Synonym 2-methylsulfonyl ethanol, 2-hydroxyethyl methyl sulfone, 2-methanesulfonylethanol, 2-methanesulfonylethan-1-ol, 2-methanesulfonyl-ethanol, 2-methanesulfonyl ethanol, ethanol, 2-methylsulfonyl, unii-5fi8eb2k0l, 2-methylsulphonyl ethanol, 2-methylsulfonyl ethyl alcohol
Solubility Information Soluble in water.
InChI Key KFTYFTKODBWKOU-UHFFFAOYSA-N
SMILES CS(=O)(=O)CCO
IUPAC Name 2-methylsulfonylethanol
Molecular Weight (g/mol) 124.154
PubChem CID 84834
Formula Weight 124.16
Percent Purity 97%
Chemical Name or Material 2-(Methylsulfonyl)ethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.