missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Methylsulfonyl)acetonitrile, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 83.0°C to 86.0°C |
Color | White |
Elemental Analysis | 29.03 to 31.45% C, 11.29 to 12.23% N |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 5 g |
Formula Weight | 119.14 |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- 2-(Methylsulfonyl)acetonitrile
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'