missing translation for 'onlineSavingsMsg'
Learn More

2-Methylsulfonyl-5-(tri-n-butylstannyl)pyrimidine, 96%, Thermo Scientific™

Product Code. 15409235
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15409235 1 g 1g
15419235 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409235 Supplier Thermo Scientific Alfa Aesar Supplier No. H51574.03
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122476-85-1
Molecular Formula C17H32N2O2SSn
Molecular Weight (g/mol) 447.225
MDL Number MFCD07787379
InChI Key RRHTVQOGUAOHDR-UHFFFAOYSA-N
Synonym 2-methylsulfonyl-5-tributylstannyl pyrimidine, 2-methanesulfonyl-5-tributylstannyl pyrimidine, 2-methylsulphonyl-5-tributylstannyl pyrimidine, tributyl-2-methylsulfonylpyrimidin-5-yl stannane, 2-methylsulfonyl-5-tri-n-butylstannyl pyrimidine
PubChem CID 2763272
IUPAC Name tributyl-(2-methylsulfonylpyrimidin-5-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CN=C(N=C1)S(=O)(=O)C

Specifications

Quantity 1 g
UN Number UN2788
Formula Weight 447.22
Percent Purity 96%
Chemical Name or Material 2-Methylsulfonyl-5-(tri-n-butylstannyl)pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.