missing translation for 'onlineSavingsMsg'
Learn More

2-Methylsuccinic acid, MedChemExpress

Product Code. 16423429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16423429 Supplier Medchem Express Supplier No. HYW010381

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 2-Methylsuccinic acid is a normal metabolite in human fluids and the main biochemical measurable features in ethylmalonic encephalopathy.

2-Methylsuccinic acid is a normal metabolite in human fluids and the main biochemical measurable features in ethylmalonic encephalopathy.

Specifications

Chemical Name or Material 2-Methylsuccinic acid
CAS 498-21-5
Color White
Purity Grade Notes Research
Molecular Formula C5H8O4
Quantity 1 g
Solubility Information DMSO : ≥ 100 mg/mL (756.89 mM)
SMILES O=C(O)C(C)CC(O)=O
Molecular Weight (g/mol) 132.12
Formula Weight 132.12
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.