Learn More
2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
Description
2-Methylstyrene is a monomer used in the preparation of poly(2-methylstyrene). It is also used as an intermediate in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Methylstyrene is a monomer used in the preparation of poly(2-methylstyrene). It is also used as an intermediate in synthetic chemistry.
Solubility
Slightly miscible with water.
Notes
Store in a cool place. Incompatible with oxidizing agents and strong acids.
Specifications
Specifications
| Melting Point | -69°C |
| Density | 0.916 |
| Boiling Point | 170°C to 172°C |
| Flash Point | 58°C (136°F) |
| Refractive Index | 1.545 |
| Quantity | 5 g |
| UN Number | UN2618 |
| Beilstein | 1901764 |
| Solubility Information | Slightly miscible with water. |
| Formula Weight | 118.18 |
| Show More |
Safety and Handling
- 2-Methylstyrene
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contains : 2-Methylstyrene
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.