missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylstyrene, 97%, stabilized
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
Unit Size:
5mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS | 611-15-4 |
| Melting Point | -69°C |
| Density | 0.9140g/mL |
| Boiling Point | 169°C to 171°C |
| Flash Point | 58°C |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Molecular Formula | C9H10 |
| Quantity | 5 mL |
| Specific Gravity | 0.914 |
| Show More |
Safety and Handling
Product Identifier
- 2-Methylstyrene
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- H411-Toxic to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction