missing translation for 'onlineSavingsMsg'
Learn More

2-Methylquinoxaline, Dr. Ehrenstorfer

Product Code. 30327243
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327243 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327243 Supplier Dr. Ehrenstorfer Supplier No. DREC15083220
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methylquinoxaline
CAS 7251-61-8
Analyte or Component Names 2-Methylquinoxaline
Shipping Condition +20°C
Molecular Formula C9 H8 N2
Certifications/Compliance ISO/IEC 17025
Synonym NSC 65587
InChI Formula InChI=1S/C9H8N2/c1-7-6-10-8-4-2-3-5-9(8)11-7/h2-6H,1H3
SMILES Cc1cnc2ccccc2n1
IUPAC Name 2-methylquinoxaline
Molecular Weight (g/mol) 144.1732
Quantity 250 mg
Formula Weight 144.0687 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.