missing translation for 'onlineSavingsMsg'
Learn More

2-Methylquinoxaline, 95%

Product Code. 11452618
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452618 10 g 10g
11462618 50 g 50g
11472618 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452618 Supplier Thermo Scientific Alfa Aesar Supplier No. A15895.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7251-61-8 | C9H8N2 | 144.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7251-61-8
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00006727
InChI Key ALHUXMDEZNLFTA-UHFFFAOYSA-N
Synonym quinoxaline, 2-methyl, 2-methyl quinoxaline, unii-03vu31mv6j, ccris 2948, methyl-quinoxaline, 2-methyl-quinoxaline, acmc-1biyr, 2-methylbenzopyrazine, 2-methyl-1,4-naphthyridine, 5-23-07-00219 beilstein handbook reference
PubChem CID 23686
IUPAC Name 2-methylquinoxaline
SMILES CC1=NC2=CC=CC=C2N=C1

Specifications

Density 1.118
Boiling Point 245°C to 247°C
Flash Point 107°C (224°F)
Refractive Index 1.617
Quantity 10 g
Beilstein 113307
Formula Weight 144.18
Percent Purity 97%
Chemical Name or Material 2-Methylquinoxaline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.