missing translation for 'onlineSavingsMsg'
Learn More

2-Methylquinoxaline, 97%

Product Code. 10285030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285030 100 g Glass bottle 100g
10351134 5 g Glass bottle 5g
10735051 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10285030 Supplier Thermo Scientific Acros Supplier No. 127661000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7251-61-8 | C9H8N2 | 144.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7251-61-8
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00006727
InChI Key ALHUXMDEZNLFTA-UHFFFAOYSA-N
Synonym quinoxaline, 2-methyl, 2-methyl quinoxaline, unii-03vu31mv6j, ccris 2948, methyl-quinoxaline, 2-methyl-quinoxaline, acmc-1biyr, 2-methylbenzopyrazine, 2-methyl-1,4-naphthyridine, 5-23-07-00219 beilstein handbook reference
PubChem CID 23686
IUPAC Name 2-methylquinoxaline
SMILES CC1=NC2=CC=CC=C2N=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Orange to Red
Density 1.1100g/mL
Boiling Point 245°C to 247°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6120 to 1.617
Quantity 100 g
Beilstein 23, I, 44
Specific Gravity 1.11
Solubility Information Solubility in water: miscible. Other solubilities: miscible in ether,acetone,benzene,very soluble in ethanol,soluble in carbon tetrachloride
Formula Weight 144.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methylquinoxaline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylquinoxaline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.