missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrrolidine, 97%

Product Code. 11398297
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398297 1 g 1g
11308307 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398297 Supplier Thermo Scientific Alfa Aesar Supplier No. 031902.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 765-38-8 | C5H11N | 85.15 g/mol

2-Methylpyrrolidine can be used to produce 2-(2-methylpyrrolidinyl)benzaldehyde at the temperature of 152°C. It is commonly used as organic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-38-8
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00014491
InChI Key RGHPCLZJAFCTIK-UHFFFAOYSA-N
PubChem CID 13003
IUPAC Name 2-methylpyrrolidine
SMILES CC1CCCN1

Specifications

Density 0.834 g/mL
Flash Point 2°C (37°F)
Odor Amine-like
Refractive Index 1.437
Quantity 1 g
UN Number UN2733
Synonym pyrrolidine, 2-methyl, 2-methyl-pyrrolidine, 2-methylpyrolidine, 2-methyl-pyrolidine, 2-methyl pyrrolidine, pubchem9410, pyrrolidine,2-methyl, acmc-209mch, acmc-1aspi, rac-2-methyl-pyrrolidine
Solubility Information Soluble in water.
Formula Weight 85.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methylpyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.