missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylpyrrolidine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Methylpyrrolidine can be used to produce 2-(2-methylpyrrolidinyl)benzaldehyde at the temperature of 152°C. It is commonly used as organic reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.834 g/mL |
Flash Point | 2°C (37°F) |
Odor | Amine-like |
Refractive Index | 1.437 |
Quantity | 5 g |
UN Number | UN2733 |
Synonym | pyrrolidine, 2-methyl, 2-methyl-pyrrolidine, 2-methylpyrolidine, 2-methyl-pyrolidine, 2-methyl pyrrolidine, pubchem9410, pyrrolidine,2-methyl, acmc-209mch, acmc-1aspi, rac-2-methyl-pyrrolidine |
Solubility Information | Soluble in water. |
Formula Weight | 85.15 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'