missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrimidine, 98%

Product Code. 15539503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15539503 5 g 5g
15529503 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15539503 Supplier Thermo Scientific Acros Supplier No. 459860050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5053-43-0 | C5H6N2 | 94.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5053-43-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.11
InChI Key LNJMHEJAYSYZKK-UHFFFAOYSA-N
Synonym pyrimidine, 2-methyl, 2-methyl pyrimidine, pubchem21974, 2-methyl-1,3-diazine, 2-methylpyrimidine, pyrimidine, 2-methyl-6ci,7ci,8ci,9ci
PubChem CID 78748
IUPAC Name 2-methylpyrimidine
SMILES CC1=NC=CC=N1

Specifications

Melting Point -6.0°C
Density 1.0132g/mL
Boiling Point 130.0°C to 132.0°C
Flash Point 37.8°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 1.0132
Formula Weight 94.11
Percent Purity 98%
Chemical Name or Material 2-Methylpyrimidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methylpyrimidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.