missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrazine, 99+%

Product Code. 10284790 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10284790 100 g Glass bottle 100g
10571371 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10284790 Supplier Thermo Scientific Acros Supplier No. 127551000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 109-08-0 | C5H6N2 | 94.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-08-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00006142
InChI Key CAWHJQAVHZEVTJ-UHFFFAOYSA-N
Synonym methylpyrazine, pyrazine, methyl, pyrazine, 2-methyl, 2-methyl-1,4-diazine, 2-methyl pyrazine, 2-methyl-pyrazine, unii-rvc6500u9c, fema no. 3309, ccris 2927, wln: t6n dnj b1
PubChem CID 7976
IUPAC Name 2-methylpyrazine
SMILES CC1=NC=CN=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -29.0°C
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 134.0°C to 137.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5042
Quantity 100 g
Beilstein 23, 94
Specific Gravity 1.03
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols, ether, acetone, oils and, halogenated solvents
Formula Weight 94.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Methylpyrazine, 99+ %
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methylpyrazine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.