missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyrazine-5-carboxylic acid, 98%, Thermo Scientific™

Product Code. 11479070
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479070 25 g 25g
11459070 1 g 1g
11469070 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479070 Supplier Thermo Scientific Alfa Aesar Supplier No. A14011.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5521-55-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00068241
InChI Key RBYJWCRKFLGNDB-UHFFFAOYSA-N
Synonym 5-methyl-2-pyrazinecarboxylic acid, 2-methylpyrazine-5-carboxylic acid, pyrazinecarboxylic acid, 5-methyl, unii-b861rs5nhi, 5-methylpyrazinecarboxylic acid, 5-methyl pyrazine-2-carboxylic acid, 5-methyl-pyrazine-2-carboxylic acid, 2-carboxy-5-methylpyrazine, 2-methyl-5-pyrazinoic acid, b861rs5nhi
PubChem CID 122831
IUPAC Name 5-methylpyrazine-2-carboxylic acid
SMILES CC1=NC=C(N=C1)C(=O)O

Specifications

Melting Point 165°C to 167°C
Quantity 25 g
Beilstein 116005
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 2-Methylpyrazine-5-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.