missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpropylboronic Acid, TRC

Product Code. 30480654
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480654 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480654 Supplier Toronto Research Chemicals Supplier No. TRCM3267451G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 84110-40-7
Molecular Formula C4H11BO2
Quantity 1 g
Synonym B-(2-methylpropyl)boronic Acid, 2-Methyl-1-propaneboronic Acid, (2-Methylpropyl)boronic Acid, Isobutyl Boronic Acid, Isobutylboric Acid
InChI Formula InChI=1S/C4H11BO2/c1-4(2)3-5(6)7/h4,6-7H,3H2,1-2H3
SMILES CC(C)CB(O)O
IUPAC Name 2-methylpropylboronic acid
Molecular Weight (g/mol) 101.94
Formula Weight 102.0852
Chemical Name or Material 2-Methylpropylboronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.