missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpropene, SPEX CertiPrep™

Product Code. 15256206 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15256206 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15256206 Supplier SPEX Certiprep Supplier No. S2556

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 115-11-7
Density 0.7898 g/cm3
Odor Characteristic
Molecular Formula C4H8
MDL Number MFCD00008898
Quantity 1 mL
Synonym isobutene, isobutylene, 2-methylpropene, 2-methylprop-1-ene, 2-methylpropylene, 1-propene, 2-methyl, 2-methyl-1-propene, 1,1-dimethylethylene, isopropylidenemethylene, methylpropene
InChI Key VQTUBCCKSQIDNK-UHFFFAOYSA-N
SMILES CC(C)=C
IUPAC Name 2-methylprop-1-ene
Molecular Weight (g/mol) 56.11
PubChem CID 8255
ChEBI CHEBI:43907
Vapor Pressure 128hPa at 20°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylpropene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.