missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpropanimidamide hydrochloride, ≥95%, Thermo Scientific™

Product Code. p-2698659
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
50g
Product Code. Quantity Packaging unitSize
10407822 1 g Amber Glass Bottle 1g
10541132 10 g Amber Glass Bottle 10g
10059381 50 g Amber Glass Bottle 50g
3 options
This item is not returnable. View return policy

Product Code. 10407822

Brand: Thermo Scientific Maybridge RF02291DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22007-68-7
Molecular Formula C4H11ClN2
Molecular Weight (g/mol) 122.596
MDL Number MFCD00173787
InChI Key VWXLCWNPSOUPPE-UHFFFAOYSA-N
Synonym 2-methylpropanimidamide hydrochloride, isobutyrimidamide hydrochloride, isobutyramidine hydrochloride, 2-methylpropionamidine hcl, isobutanimidamide hydrochloride, isopropylcarbamidine hydrochloride, 2-methylpropionamidine hydrochloride, 2-methyl propanimidamide hydrochloride, isobutanimidamide hcl, sothiadhk\up@
PubChem CID 2782053
IUPAC Name 2-methylpropanimidamide;hydrochloride
SMILES CC(C)C(=N)N.Cl

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Linear Formula (CH3)2CHC(=NH)NH2·HCl
Quantity 1 g
Formula Weight 122.6
Percent Purity 90%
Chemical Name or Material 2-Methylpropanimidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.