missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpiperidine, 98+%

Product Code. 11357745
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357745 100 mL 100mL
11367745 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357745 Supplier Thermo Scientific Alfa Aesar Supplier No. L02567.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 109-05-7 | C6H13N | 99.18 g/mol

2-Methylpiperidine, is used for the oxidation of secondary amines to nitrones by means of H2O2 catalyzed by Sodium tungsten oxide dihydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylpiperidine, is used for the oxidation of secondary amines to nitrones by means of H2O2 catalyzed by Sodium tungsten oxide dihydrate.

Solubility
It is easily soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-05-7
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
MDL Number MFCD00005982
InChI Key NNWUEBIEOFQMSS-UHFFFAOYNA-N
Synonym 2-pipecoline, pipicoline, alpha-pipecoline, piperidine, 2-methyl, alpha-pipecolin, pipecoline, alpha, 2-methyl-piperidine, alpha-methylpiperidine, .alpha.-pipecolin, .alpha.-pipecoline
PubChem CID 7974
IUPAC Name 2-methylpiperidine
SMILES CC1CCCCN1

Specifications

Melting Point -4°C
Density 0.842
Boiling Point 118°C to 119°C
Flash Point 8°C (46°F)
Odor Amine-like
Refractive Index 1.447
Quantity 100 mL
UN Number UN2733
Beilstein 79804
Solubility Information It is easily soluble in water.
Formula Weight 99.18
Percent Purity ≥98%
Chemical Name or Material 2-Methylpiperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.