missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpiperazine, TRC

Product Code. 17983785
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983785 50 g 50g
17961615 1 g 1g
17932925 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17983785 Supplier Toronto Research Chemicals Supplier No. TRCM32480550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-07-9
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.16
Synonym 2-Methyl-piperazine, (±)-2-Methyl-piperazine, (±)-2-Methylpiperazine, NSC 5277
IUPAC Name 2-methylpiperazine
SMILES CC1CNCCN1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C5H12N2/c1-5-4-6-2-3-7-5/h5-7H,2-4H2,1H3
Formula Weight 100.1
Chemical Name or Material 2-Methylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.