missing translation for 'onlineSavingsMsg'
Learn More

(2-Methylphenyl)methanesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10336804
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336804 250 mg Amber Glass Bottle 250mg
10467023 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336804 Supplier Thermo Scientific Maybridge Supplier No. MO07798CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 92614-55-6
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD02683108
InChI Key NDMREUGSHNSYBI-UHFFFAOYSA-N
Synonym 2-methylphenyl methanesulfonyl chloride, o-tolylmethanesulfonyl chloride, o-tolyl-methanesulfonyl chloride, o-tolyl methanesulfonyl chloride, o-tolylmethanesulfonylchloride, 2-methylbenzylsulphonyl chloride, 2-methylphenyl methanesulphonyl chloride, 2-methylphenyl methyl sulfonyl chloride, benzenemethanesulfonyl chloride, 2-methyl, chloro 2-methylphenyl methyl sulfone
PubChem CID 2760661
IUPAC Name (2-methylphenyl)methanesulfonyl chloride
SMILES CC1=CC=CC=C1CS(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material (2-Methylphenyl)methanesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.