missing translation for 'onlineSavingsMsg'
Learn More

2-Methylphenanthrene, TRC

Product Code. 30138159
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138159 250 mg 250mg
17952495 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138159 Supplier Toronto Research Chemicals Supplier No. TRCM325420250MG
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2531-84-2
Molecular Formula C15 H12
Molecular Weight (g/mol) 192.26
Synonym 2-Methylphenanthrene, Phenanthrene, 2-methyl- (8CI, 9CI, ACI), 2-Methylphenanthrene (ACI), NSC 89276
IUPAC Name 2-methylphenanthrene
SMILES Cc1ccc2c(ccc3ccccc23)c1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H12/c1-11-6-9-15-13(10-11)8-7-12-4-2-3-5-14(12)15/h2-10H,1H3
Formula Weight 192.0939
Percent Purity >95%
Chemical Name or Material 2-Methylphenanthrene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.