Product Code. 17951855
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
17951855
25 mL
25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
17951855
Supplier
Toronto Research Chemicals
Supplier No.
TRCM22638025ML
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Specifications
CAS
107-83-5
Molecular Formula
C6 H14
Quantity
25 mL
Synonym
Pentane, 2-methyl-, 2-Methylpentane, 1,1-Dimethylbutane, Isohexane, Kyowasol C 600M, Kyowazol C 600, NSC 66496
InChI Formula
InChI=1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3
SMILES
CCCC(C)C
IUPAC Name
2-methylpentane
Molecular Weight (g/mol)
86.18
Formula Weight
86.1096
Chemical Name or Material
2-Methylpentane
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?