missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnonane, 98+%

Product Code. 11475796
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11475796

Brand: Thermo Scientific Alfa Aesar B20849.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 871-83-0 | C10H22 | 142.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871-83-0
Molecular Formula C10H22
Molecular Weight (g/mol) 142.29
MDL Number MFCD00027321
InChI Key SGVYKUFIHHTIFL-UHFFFAOYSA-N
Synonym nonane, 2-methyl, c9-11 isoparaffin, alkanes, c9-11-iso, 2-methylnonane, unii-382akh052v, isodecane, octane, dimethyl, iso-decane, 2-methyl nonane, c10-13 isoparaffin
PubChem CID 13379
IUPAC Name 2-methylnonane
SMILES CCCCCCCC(C)C

Specifications

Density 0.72
Boiling Point 166°C
Flash Point 46°C (115°F)
Odor Petroleum-like
Linear Formula CH3(CH2)6CH(CH3)2
Refractive Index 1.41
Quantity 25 g
UN Number UN3295
Beilstein 1696990
Formula Weight 142.29
Percent Purity ≥98%
Chemical Name or Material 2-Methylnonane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.