missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnicotinic acid, 98+%, Thermo Scientific Chemicals

Catalog Number 10061594
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3222-56-8
C7H7NO2
137.14
MFCD00013449
HNTZKNJGAFJMHQ-UHFFFAOYSA-N
2-methylnicotinic acid, 2-methyl-nicotinic acid, 2-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-methyl, 2-methyl nicotinic acid, 2-methyl-3-pyridinecarboxylic acid, methylnicotinic acid, pubchem2458, 2-methylnicotinoic acid, acmc-1ctxp
643373
2-methylpyridine-3-carboxylic acid
CC1=C(C=CC=N1)C(=O)O
This item is not returnable. View return policy
3222-56-8
C7H7NO2
137.14
MFCD00013449
HNTZKNJGAFJMHQ-UHFFFAOYSA-N
2-methylnicotinic acid, 2-methyl-nicotinic acid, 2-methylnicotinicacid, 3-pyridinecarboxylic acid, 2-methyl, 2-methyl nicotinic acid, 2-methyl-3-pyridinecarboxylic acid, methylnicotinic acid, pubchem2458, 2-methylnicotinoic acid, acmc-1ctxp
643373
2-methylpyridine-3-carboxylic acid
CC1=C(C=CC=N1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 225.0°C to 228.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 137.14
Percent Purity 98+%
Physical Form Crystals or Powder
Chemical Name or Material 2-Methylnicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylnicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'