missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnaphthalene, 96%

Product Code. 10473395 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10473395 2.5 kg Glass bottle 2.5kg
10031170 5 g Glass bottle 5g
10647092 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10473395 Supplier Thermo Scientific Acros Supplier No. 127170025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-57-6 | C11H10 | 142.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-57-6
Molecular Formula C11H10
Molecular Weight (g/mol) 142.2
MDL Number MFCD00004118
InChI Key QIMMUPPBPVKWKM-UHFFFAOYSA-N
Synonym naphthalene, 2-methyl, beta-methylnaphthalene, .beta.-methylnaphthalene, naphthalene, beta-methyl, unii-s8mcx3c16h, 2-methyl naphthalene, 2-methyl-naphthalene, beta-methyl naphthalenes, s8mcx3c16h, naphtalene,2-methyl mfc11 h10
PubChem CID 7055
ChEBI CHEBI:50720
IUPAC Name 2-methylnaphthalene
SMILES CC1=CC2=CC=CC=C2C=C1

Specifications

CAS Min % 96
Melting Point 32°C to 36°C
Color White
Density 1.0100g/mL
Boiling Point 241°C to 242°C
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH3
Quantity 2.5 kg
Beilstein 05, 567
Specific Gravity 1.01
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 142.2
Percent Purity 96%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2-Methylnaphthalene, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.