missing translation for 'onlineSavingsMsg'
Learn More

2-Methylisoindole-1,3-dione, TRC

Product Code. 30468410
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468410 25 g 25g
30472599 5 g 5g
30464928 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30468410 Supplier Toronto Research Chemicals Supplier No. TRCM31436025G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 550-44-7
Molecular Formula C9 H7 N O2
Molecular Weight (g/mol) 161.16
Synonym 2-Methyl-1H-isoindole-1,3(2H)-dione, Phthalimide, N-methyl- (6CI,7CI,8CI), 2-Methyl-1H-isoindole-1,3(2H)-dione, 2-Methylisoindole-1,3-dione, 2-Methylisoindoline-1,3-dione, N-Methylphthalimide, NSC 44059
IUPAC Name 2-methylisoindole-1,3-dione
SMILES CN1C(=O)c2ccccc2C1=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H7NO2/c1-10-8(11)6-4-2-3-5-7(6)9(10)12/h2-5H,1H3
Formula Weight 161.0477
Chemical Name or Material Phthalimide-N-methyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.