missing translation for 'onlineSavingsMsg'
Learn More

2-Methylindoline, 99+%, Thermo Scientific Chemicals

Product Code. 10636471
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6872-06-6
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00005706
InChI Key QRWRJDVVXAXGBT-UHFFFAOYNA-N
Synonym 2-methylindoline, 2-methyl-2,3-dihydro-1h-indole, 2,3-dihydro-2-methylindole, 1h-indole, 2,3-dihydro-2-methyl, indoline, 2-methyl, 2-methyl-2,3-dihydroindole, dihydroindole, 2-methyl, alpha-methyldihydroindole, .alpha.-methyldihydroindole, 2,3-dihydro-2-methyl-1h-indole
PubChem CID 23305

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -51°C
Color Yellow to Brown
Density 1.0200g/mL
Boiling Point 227°C (1013.0 Mbar)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.5680 to 1.57
Quantity 5 mL
Beilstein 20, 279
Specific Gravity 1.02
Solubility Information Solubility in water: negligible
Formula Weight 133.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Methylindoline, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylindoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'