missing translation for 'onlineSavingsMsg'
Learn More

2-Methylindoline, 99+%

Product Code. 10104780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10104780 100 mL Glass bottle 100mL
10636471 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10104780 Supplier Thermo Scientific Acros Supplier No. 275881000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6872-06-6 | C9H11N | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-6-6872
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.19
MDL Number MFCD00005706
InChI Key QRWRJDVVXAXGBT-UHFFFAOYNA-N
Synonym 2-methylindoline, 2-methyl-2,3-dihydro-1h-indole, 2,3-dihydro-2-methylindole, 1h-indole, 2,3-dihydro-2-methyl, indoline, 2-methyl, 2-methyl-2,3-dihydroindole, dihydroindole, 2-methyl, alpha-methyldihydroindole, .alpha.-methyldihydroindole, 2,3-dihydro-2-methyl-1h-indole
PubChem CID 23305

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -51°C
Color Yellow to Brown
Density 1.0200g/mL
Boiling Point 227°C (1013.0 Mbar)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.5680 to 1.57
Quantity 100 mL
Beilstein 20, 279
Specific Gravity 1.02
Solubility Information Solubility in water: negligible
Formula Weight 133.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Methylindoline, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylindoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.