missing translation for 'onlineSavingsMsg'
Learn More

2-Methylindole, 98+%

Product Code. 11439662
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439662 1000 g 1000g
11449662 50 g 50g
11459662 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439662 Supplier Thermo Scientific Alfa Aesar Supplier No. A10764.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-20-5 | C9H9N | 131.178 g/mol

2-Methylindole is used as a reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction, Friedel-Crafts alkylation reactions, preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, Michael addition reactions and in synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylindole is used as a reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction, Friedel-Crafts alkylation reactions, preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, Michael addition reactions and in synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-20-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005616
InChI Key BHNHHSOHWZKFOX-UHFFFAOYSA-N
Synonym 2-methylindole, 1h-indole, 2-methyl, indole, 2-methyl, 2-methyl indole, unii-i7cn58827i, 2-methylindol, methylketole, methyl indole, 2-methyl-indole, pubchem7504
PubChem CID 7224
ChEBI CHEBI:49402
IUPAC Name 2-methyl-1H-indole
SMILES CC1=CC2=CC=CC=C2N1

Specifications

Melting Point 56°C to 60°C
Density 1.07
Boiling Point 272°C to 273°C
Flash Point 141°C (285°F)
Quantity 1000 g
Beilstein 109781
Sensitivity Air and light sensitive
Solubility Information Insoluble in water.
Formula Weight 131.18
Percent Purity ≥98%
Chemical Name or Material 2-Methylindole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.