missing translation for 'onlineSavingsMsg'
Learn More

2-Methylindole-3-carboxaldehyde, 97+%, Thermo Scientific Chemicals

Product Code. 10381413 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10381413 25 g Glass bottle 25g
10391413 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10381413 Supplier Thermo Scientific Acros Supplier No. 379720250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5416-80-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00012077
InChI Key CYZIVXOEJNAIBS-UHFFFAOYSA-N
Synonym 2-methylindole-3-carboxaldehyde, 3-formyl-2-methylindole, 2-methyl-3-formylindole, 1h-indole-3-carboxaldehyde, 2-methyl, 2-methylindole-3-carbaldehyde, indole-3-carboxaldehyde, 2-methyl, pubchem7437, zlchem 1339, acmc-1apkw, ksc271e8j
PubChem CID 73166
IUPAC Name 2-methyl-1H-indole-3-carbaldehyde
SMILES CC1=C(C2=CC=CC=C2N1)C=O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 204°C to 205°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 159.19
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 2-Methylindole-3-carboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.