missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazole, 97%

Product Code. 11448926
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448926 2500 g 2500g
11428926 100 g 100g
11438926 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448926 Supplier Thermo Scientific Alfa Aesar Supplier No. A17063.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 693-98-1 | C4H6N2 | 82.11 g/mol

2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics, which is useful in combat anaerobic bacterial and parasitic infections. It acts as a ligand in coordination chemistry. It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes. Further, it acts as a catalyst for refolding of enhanced colored fluorescent protein. It is also useful as a building block in the synthesis of biologically active compounds. In addition to this, it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics, which is useful in combat anaerobic bacterial and parasitic infections. It acts as a ligand in coordination chemistry. It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes. Further, it acts as a catalyst for refolding of enhanced colored fluorescent protein. It is also useful as a building block in the synthesis of biologically active compounds. In addition to this, it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole.

Solubility
Soluble in water and ethanol.

Notes
Incompatible with strong oxidizing agents, acids, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-98-1
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005190
InChI Key LXBGSDVWAMZHDD-UHFFFAOYSA-N
Synonym 2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107
PubChem CID 12749
IUPAC Name 2-methyl-1H-imidazole
SMILES CC1=NC=CN1

Specifications

Melting Point 142°C to 144°C
Boiling Point 267°C to 268°C
Flash Point 155°C (311°F)
Quantity 2500 g
UN Number UN3259
Beilstein 1368
Solubility Information Soluble in water and ethanol.
Formula Weight 82.11
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 2-Methylimidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.