missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexanoyl chloride, 97%

Product Code. 11334806
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334806 1 g 1g
11324806 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334806 Supplier Thermo Scientific Alfa Aesar Supplier No. H32829.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 41693-47-4 | C7H13ClO | 148.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41693-47-4
Molecular Formula C7H13ClO
Molecular Weight (g/mol) 148.63
MDL Number MFCD01318644
InChI Key WKEBLSOTVYFYKY-UHFFFAOYNA-N
Synonym 2-methylcaproyl chloride, 2-methyl hexanoyl chloride, hexanoyl chloride,2-methyl
PubChem CID 3458543
IUPAC Name 2-methylhexanoyl chloride
SMILES CCCCC(C)C(Cl)=O

Specifications

Density 0.957
Boiling Point 163°C
Flash Point 55°C (131°F)
Refractive Index 1.429
Quantity 1 g
UN Number UN2920
Sensitivity Moisture sensitive
Formula Weight 148.63
Percent Purity 97%
Chemical Name or Material 2-Methylhexanoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.