missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexanoic acid, 98%

Product Code. 11483664
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483664 100 g 100g
11463664 5 g 5g
11473664 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483664 Supplier Thermo Scientific Alfa Aesar Supplier No. A11290.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4536-23-6 | C7H13O2 | 129.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4536-23-6
Molecular Formula C7H13O2
Molecular Weight (g/mol) 129.18
MDL Number MFCD00002674
InChI Key CVKMFSAVYPAZTQ-ZCFIWIBFSA-M
Synonym 2-methylcaproic acid, hexanoic acid, 2-methyl, methylhexanoic acid, 2-methyl hexanoic acid, 2-hexanecarboxylic acid, alpha-methylcaproic acid, 2-methyl-hexanoic acid, fema no. 3191, .alpha.-methylcaproic acid, hexanoic acid, methyl
PubChem CID 20653
SMILES CCCC[C@@H](C)C([O-])=O

Specifications

Density 0.916
Boiling Point 209°C to 210°C
Flash Point 105°C (221°F)
Odor Rancid
Refractive Index 1.4205
Quantity 100 g
UN Number UN3265
Beilstein 1721227
IUPAC Name 2-methylhexanoic acid
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 2-Methylhexanoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.