missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhex-2-enoic Acid, TRC

Product Code. 30380382
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380382 1 g
30380619 100 mg
30379496 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380382 Supplier Toronto Research Chemicals Supplier No. TRCE5215851G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28897-58-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
IUPAC Name (E)-2-methylhex-2-enoic acid
SMILES CCC\C=C(/C)\C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H12O2/c1-3-4-5-6(2)7(8)9/h5H,3-4H2,1-2H3,(H,8,9)/b6-5+
Formula Weight 128.0837
Chemical Name or Material 2-Methylhex-2-enoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.