missing translation for 'onlineSavingsMsg'
Learn More

2-Methylheptanoic acid, 98%

Product Code. p-7026190
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11429547

Brand: Thermo Scientific Alfa Aesar B22050.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1188-02-9 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1188-02-9
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00142944
InChI Key NKBWMBRPILTCRD-UHFFFAOYSA-N
Synonym 2-methylenanthic acid, heptanoic acid, 2-methyl, methylamylacetic acid, 2-methyl-heptanoic acid, heptanoic acid, methyl, alpha-methylheptanoic acid, 2-methyl heptanoic acid, heptane-2-carboxylic acid, fema no. 2706, heptanoic acid, 2-methyl-, 2s
PubChem CID 14475
IUPAC Name 2-methylheptanoic acid
SMILES CCCCCC(C)C(=O)O

Specifications

Density 0.906
Boiling Point 139°C to 141°C (30 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4255
Quantity 5 g
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material 2-Methylheptanoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.