missing translation for 'onlineSavingsMsg'
Learn More

2-Methylheptanoic acid, 98%

Product Code. 11449547
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449547 100 g 100g
11429547 5 g 5g
11439547 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449547 Supplier Thermo Scientific Alfa Aesar Supplier No. B22050.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1188-02-9 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1188-02-9
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00142944
InChI Key NKBWMBRPILTCRD-UHFFFAOYSA-N
Synonym 2-methylenanthic acid, heptanoic acid, 2-methyl, methylamylacetic acid, 2-methyl-heptanoic acid, heptanoic acid, methyl, alpha-methylheptanoic acid, 2-methyl heptanoic acid, heptane-2-carboxylic acid, fema no. 2706, heptanoic acid, 2-methyl-, 2s
PubChem CID 14475
IUPAC Name 2-methylheptanoic acid
SMILES CCCCCC(C)C(=O)O

Specifications

Density 0.906
Boiling Point 139°C to 141°C (30 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4255
Quantity 100 g
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material 2-Methylheptanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.