missing translation for 'onlineSavingsMsg'
Learn More

2-Methylheptane, 99%

Product Code. 10056183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10056183 1 g Glass bottle 1g
10347150 5 g Glass bottle 5g
10572711 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10056183 Supplier Thermo Scientific Acros Supplier No. 126880010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 592-27-8 | C8H18 | 114.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-27-8
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00009518
InChI Key JVSWJIKNEAIKJW-UHFFFAOYSA-N
Synonym heptane, 2-methyl, methylheptane, 2-methyl-heptane, unii-yu6su8ccvb, yu6su8ccvb, hexane, dimethyl, dimethylhexane, acmc-1avkm, 2-methylheptane, ch3 2ch ch2 4ch3
PubChem CID 11594
IUPAC Name 2-methylheptane
SMILES CCCCCC(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -108.9°C
Color Colorless
Density 0.6900g/mL
Boiling Point 116°C (761.0 mmHg)
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH(CH3)2
Refractive Index 1.3940 to 1.396
Quantity 1 g
Beilstein 01, 161
Specific Gravity 0.69
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,miscible with ethanol,acetone
Formula Weight 114.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylheptane, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylheptane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.