missing translation for 'onlineSavingsMsg'
Learn More

2-Methylglutaric Acid, TRC

Product Code. 30479187
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479187 5 g 5g
30468593 10 g 10g
30476341 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479187 Supplier Toronto Research Chemicals Supplier No. TRCM2200985G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-62-9
Molecular Formula C6 H10 O4
Molecular Weight (g/mol) 146.14
Synonym 2-Methylpentanedioic Acid, (±)-2-Methylglutaric Acid, 2-Methylglutaric Acid, NSC 408456, NSC 5941, Pentanedioic Acid 2-Methyl Ester, α-Methylglutaric Acid
IUPAC Name 2-methylpentanedioic acid
SMILES CC(CCC(=O)O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H10O4/c1-4(6(9)10)2-3-5(7)8/h4H,2-3H2,1H3,(H,7,8)(H,9,10)
Formula Weight 146.0579
Chemical Name or Material 2-Methylglutaric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.