missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylglutaric acid, 98%, Thermo Scientific Chemicals
27.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Incompatible with bases and oxidizing agents.
2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Incompatible with bases and oxidizing agents.
Specifications
Melting Point | 74°C to 78°C |
Boiling Point | 214°C to 215°C (22 mmHg) |
Quantity | 5 g |
Beilstein | 1723219 |
Solubility Information | Soluble in water. |
Formula Weight | 146.14 |
Percent Purity | 98% |
Chemical Name or Material | 2-Methylglutaric acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'