missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclopropanecarboxylic acid, cis + trans, 96%

Product Code. 11377379
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377379 10 g 10g
11367379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377379 Supplier Thermo Scientific Alfa Aesar Supplier No. H27815.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29555-02-0 | C5H7O2 | 99.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29555-02-0
Molecular Formula C5H7O2
Molecular Weight (g/mol) 99.11
MDL Number MFCD00001293
InChI Key AYEGPMGNMOIHDL-IUYQGCFVSA-M
Synonym 2-methylcyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 2-methyl, 2-methyl-1-cyclopropanecarboxylic acid, trans-2-methylcyclopropanecarboxylic acid, cis-2-methylcyclopropanecarboxylic acid, acmc-1cg8e, 2-methyl-cyclopropanecarboxylic acid, 2-methyl cyclopropane carboxylic acid, cyclopropanecarboxylicacid, 2-methyl
PubChem CID 99873
IUPAC Name 2-methylcyclopropane-1-carboxylic acid
SMILES C[C@H]1C[C@H]1C([O-])=O

Specifications

Density 1.027
Boiling Point 200°C to 203°C
Flash Point 88°C (190°F)
Refractive Index 1.439
Quantity 10 g
UN Number UN3265
Beilstein 2323873
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material 2-Methylcyclopropanecarboxylic acid, Cis + Trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.