missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclopentanone, 98%

Product Code. 10772675 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10772675 25 g Glass bottle 25g
10144720 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772675 Supplier Thermo Scientific Acros Supplier No. 247660250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1120-72-5 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-72-5
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00001414
InChI Key ZIXLDMFVRPABBX-UHFFFAOYNA-N
Synonym 2-methylcyclopentanone, cyclopentanone, 2-methyl, .alpha.-methylcyclopentanone, cyclopentanone, methyl, alpha-methylcyclopentanone, cyclopentan-1-one, 2-methyl, methylcyclopentanone, 2-methyl cyclopentanone, 3-methyl-2-cyclopentanone, 2-methyl-cyclopentan-1-one
PubChem CID 14265
IUPAC Name 2-methylcyclopentan-1-one
SMILES CC1CCCC1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 139°C to 140°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4340 to 1.436
Quantity 25 g
Specific Gravity 0.91
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol,ether and acetone
Formula Weight 98.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylcyclopentanone
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylcyclopentanone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.