missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclopentanone, 98%

Product Code. 10144720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10144720 5 g Glass bottle 5g
10772675 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10144720 Supplier Thermo Scientific Acros Supplier No. 247660050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1120-72-5 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-72-5
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00001414
InChI Key ZIXLDMFVRPABBX-UHFFFAOYNA-N
Synonym 2-methylcyclopentanone, cyclopentanone, 2-methyl, .alpha.-methylcyclopentanone, cyclopentanone, methyl, alpha-methylcyclopentanone, cyclopentan-1-one, 2-methyl, methylcyclopentanone, 2-methyl cyclopentanone, 3-methyl-2-cyclopentanone, 2-methyl-cyclopentan-1-one
PubChem CID 14265
IUPAC Name 2-methylcyclopentan-1-one
SMILES CC1CCCC1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 139°C to 140°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4340 to 1.436
Quantity 5 g
Specific Gravity 0.91
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol,ether and acetone
Formula Weight 98.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylcyclopentanone
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methylcyclopentanone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.