missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclopentane-1,3-dione, 98%

Product Code. 11431668
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431668 100 g 100g
11411668 5 g 5g
11421668 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431668 Supplier Thermo Scientific Alfa Aesar Supplier No. A13936.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 765-69-5 | C6H8O2 | 112.128 g/mol

2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.

Solubility
Solubility in hot Methanol (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-69-5
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001406
InChI Key HXZILEQYFQYQCE-UHFFFAOYSA-N
Synonym 2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
PubChem CID 13005
IUPAC Name 2-methylcyclopentane-1,3-dione
SMILES CC1C(=O)CCC1=O

Specifications

Melting Point 212°C to 216°C
Quantity 100 g
Beilstein 1237255
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material 2-Methylcyclopentane-1,3-dione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.