missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylcyclohexanone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.
Solubility
Insoluble in water. Soluble in methanol and diethyl ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2-Methylcyclohexanone is used as a fragrant and flavoring agent. It can also be used for the preparation of a novel pladienolide analog scaffold. Also used as a solvent.
Solubility
Insoluble in water. Soluble in methanol and diethyl ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | -14°C |
Density | 0.925 |
Boiling Point | 162°C to 163°C |
Flash Point | 46°C (114°F) |
Assay Percent Range | 98% |
Refractive Index | 1.448 |
Quantity | 500 mL |
UN Number | UN2297 |
Beilstein | 506751 |
Solubility Information | Insoluble in water. Soluble in methanol and diethyl ether. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'