missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexanol, cis + trans, 97%

Product Code. 11475313
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475313 2500 mL 2500mL
11465313 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475313 Supplier Thermo Scientific Alfa Aesar Supplier No. A12877.0F
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 583-59-5 | C7H14O | 114.188 g/mol

2-Methylcyclohexanol is used in the preparation of acetic acid-(2-methyl-cyclohexyl ester) by reaction with acetic anhydride. Further, it is used to study the effect of organic solvents on epoxide hydrolase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexanol is used in the preparation of acetic acid-(2-methyl-cyclohexyl ester) by reaction with acetic anhydride. Further, it is used to study the effect of organic solvents on epoxide hydrolase.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-59-5
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00001443
InChI Key NDVWOBYBJYUSMF-UHFFFAOYSA-N
Synonym 2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
PubChem CID 11418
IUPAC Name 2-methylcyclohexan-1-ol
SMILES CC1CCCCC1O

Specifications

Melting Point -21°C
Density 0.929
Boiling Point 163°C to 166°C
Flash Point 58°C (136°F)
Refractive Index 1.462
Quantity 2500 mL
UN Number UN2617
Beilstein 1847535
Solubility Information Slightly miscible with water.
Formula Weight 114.19
Percent Purity 97%
Chemical Name or Material 2-Methylcyclohexanol, cis + trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.