missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexanol, 99%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog Number 10479080
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
583-59-5
C7H14O
114.19
MFCD00001443
NDVWOBYBJYUSMF-UHFFFAOYSA-N
2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
11418
2-methylcyclohexan-1-ol
CC1CCCCC1O
583-59-5
C7H14O
114.19
MFCD00001443
NDVWOBYBJYUSMF-UHFFFAOYSA-N
2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
11418
2-methylcyclohexan-1-ol
CC1CCCCC1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -38.0°C
Color Colorless
Density 0.9300g/mL
Boiling Point 163.0°C to 166.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H10OH
Refractive Index 1.4615 to 1.4635
Quantity 1 kg
Beilstein 06, 11
Fieser 08,303
Specific Gravity 0.93
Solubility Information Solubility in water: slightly soluble
Formula Weight 114.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylcyclohexanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'