missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclohexane-1,3-dione, 98+%

Product Code. 11423544
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423544 5 g 5g
11433544 25 g 25g
11339579 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423544 Supplier Thermo Scientific Alfa Aesar Supplier No. A11218.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1193-55-1 | C7H10O2 | 126.16 g/mol

2-Methylcyclohexane-1,3-dione acts as a synthetic intermediate. It is an important starting material, exemplified in a formal synthesis of acorone. It is also used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclohexane-1,3-dione acts as a synthetic intermediate. It is an important starting material, exemplified in a formal synthesis of acorone. It is also used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in methanol ( 0.1 g/mL). Insoluble in water.

Notes
Hygroscopic. Store at -20°C. Incompatible with oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-55-1
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00001587
InChI Key VSGJHHIAMHUZKF-UHFFFAOYSA-N
Synonym 2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g
PubChem CID 70945
IUPAC Name 2-methylcyclohexane-1,3-dione
SMILES CC1C(=O)CCCC1=O

Specifications

Melting Point 204°C to 208°C
Quantity 5 g
Beilstein 1281598
Sensitivity Hygroscopic
Solubility Information Soluble in methanol ( 0.1g/mL). Insoluble in water.
Formula Weight 126.16
Percent Purity ≥98%
Chemical Name or Material 2-Methylcyclohexane-1,3-dione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.