missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbutyryl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methylbutyryl Chloride |
| CAS | 57526-28-0 |
| Molecular Formula | C5H9ClO |
| Quantity | 2.5 g |
| Synonym | (DL)-2-Methylbutanoyl Chloride, (RS)-2-Methylbutanoyl Chloride, (±)-2-Methylbutyryl Chloride, 2-Ethylpropanoyl Chloride, 2-Methylbutanoyl Chloride, 2-Methylbutyric Acid Chloride, dl-2-Methylbutyryl Chloride, alpha-Methylbutyryl Chloride |
| InChI Formula | InChI=1S/C5H9ClO/c1-3-4(2)5(6)7/h4H,3H2,1-2H3 |
| SMILES | CCC(C)C(=O)Cl |
| IUPAC Name | 2-methylbutanoyl chloride |
| Molecular Weight (g/mol) | 120.8 |
| Formula Weight | 120.0342 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?